Assay Details
| Gene Symbol: Sgcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011892.3 | NP_036022.1 | 4 - 5 | 542 | 81 | |
| GenBank mRNA | AB024922.1 | - | 5 - 6 | 647 | 81 | |
| AF282901.1 | - | 4 - 5 | 542 | 81 | ||
| AK132610.1 | - | 4 - 5 | 487 | 81 | ||
| BC138960.1 | - | 4 - 5 | 434 | 8861982 | 81 | |
| BC138961.1 | - | 4 - 5 | 434 | 8861983 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgcg
Genomic Map