Assay Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007382.5 | NP_031408.1 | 1 - 2 | 246 | 100 | |
| GenBank mRNA | AK008149.1 | - | 1 - 2 | 48 | 100 | |
| AK131793.1 | - | 1 - 2 | 48 | 100 | ||
| AK146444.1 | - | 1 - 2 | 50 | 100 | ||
| CX232934.1 | - | 1 - 2 | 237 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131793.1, NM_007382.5 transcripts.
Set Membership: |
|

Acadm
Genomic Map