Assay Details
Gene Symbol: Gm5334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003648.2 | 1 | 1071 | 80 | ||
GenBank mRNA | BC049650.1 | - | 1 | 1070 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4933436H12 gene | 4933436H12Rik | -192.3kb | Sense | Chr. 7 - 68417424 - 68426133 on NCBI Build GRCm38 | |
Downstream | arrestin domain containing 4 | Arrdc4 |
2410003C09Rik, AV216361 |
117.0kb | Anti-sense | Chr. 7 - 68736994 - 68749238 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097333 | NR_003648.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097333 | BC049650.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P379395 | NR_003648.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P379395 | BC049650.1 | mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06472293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06319044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200981 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06319042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06235623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06235635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06643042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06319047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06051578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06319043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200973 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06235612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06282708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06235651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200998 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25201043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06282711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25200954 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10554128 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10554127 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700009372.mm.1 | TC0700001018.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700009372.mm.1 | NR_003648.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 676960 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1229780 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2780186 | - | - |
