Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400008983.mm.1 |
NM_001253755.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008983.mm.1 |
NM_001253755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008983.mm.1 |
NM_172605.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008983.mm.1 |
NM_172605.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008983.mm.1 |
TC1400001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008980.mm.1 |
NM_001253755.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008980.mm.1 |
NM_001253755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008980.mm.1 |
NM_172605.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008980.mm.1 |
NM_172605.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008980.mm.1 |
TC1400001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008976.mm.1 |
NM_001253755.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008976.mm.1 |
NM_001253755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008976.mm.1 |
NM_172605.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008976.mm.1 |
NM_172605.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008976.mm.1 |
TC1400001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008975.mm.1 |
NM_001253755.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008975.mm.1 |
NM_001253755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008975.mm.1 |
NM_172605.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008975.mm.1 |
NM_172605.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008975.mm.1 |
TC1400001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008981.mm.1 |
NM_001253755.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008981.mm.1 |
NM_001253755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008981.mm.1 |
NM_172605.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008981.mm.1 |
NM_172605.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008981.mm.1 |
TC1400001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008978.mm.1 |
NM_001253755.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008978.mm.1 |
NM_001253755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008978.mm.1 |
NM_172605.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008978.mm.1 |
NM_172605.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008978.mm.1 |
TC1400001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008985.mm.1 |
NM_001253755.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008985.mm.1 |
NM_001253755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008985.mm.1 |
NM_172605.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008985.mm.1 |
NM_172605.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008985.mm.1 |
TC1400001125.mm.1 |
MTA TC |