Assay Details
| Gene Symbol: Tdrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253755.1 | NP_001240684.1 | 6 - 7 | 787 | 61 | |
| NM_172605.3 | NP_766193.3 | 6 - 7 | 787 | 61 | ||
| GenBank mRNA | AK032892.1 | - | 6 - 7 | 732 | 61 | |
| AK033390.1 | - | 6 - 7 | 732 | 61 | ||
| BC066144.1 | - | 6 - 7 | 735 | 30435963 | 61 | |
More Information
Set Membership: |
|

Tdrd3
Genomic Map