Assay Details
Gene Symbol: Fam181b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021427.2 | NP_067402.2 | 1 | 1745 | 98 | |
GenBank mRNA | AB041569.1 | - | 1 | 1707 | 98 | |
AK140685.1 | - | 1 | 1698 | 98 | ||
AK141493.1 | - | 1 | 1343 | 98 | ||
BC043329.1 | - | 1 | 1690 | 5369836 | 98 | |
EF537862.1 | - | 1 | 667 | 98 | ||
EF537863.1 | - | 1 | 671 | 98 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328183 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328172 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328180 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328174 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328181 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328184 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328177 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328176 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328179 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328175 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328178 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | EF537863.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | AK140685.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | AK141493.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | BC043329.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | EF537862.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | EF537863.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | AK140685.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | AK141493.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | BC043329.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158678 | EF537862.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154281 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | EF537863.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | AK140685.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | AK141493.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | BC043329.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | EF537862.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | EF537863.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | AK140685.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | AK141493.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | BC043329.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158678 | EF537862.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328158 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328153 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328183 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328148 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328151 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328142 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328165 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328149 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328143 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328154 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328192 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25328185 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154281 | - | - |
Whole Mouse Genome G4122A | A_51_P158678 | EF537863.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | AK140685.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | AK141493.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | BC043329.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | EF537862.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | EF537863.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | AK140685.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | AK141493.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | BC043329.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P158678 | EF537862.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105900 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154281 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05955366 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06417671 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05955362 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328175 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328180 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328179 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328181 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328174 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328176 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328172 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328178 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328177 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328183 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25328184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25328231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328202 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05955362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06489071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06114833 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05954821 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06234914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06417671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328132 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06234908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328187 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06234906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06295049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328201 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25328140 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10565581 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10554961 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10554962 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10565580 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1442800_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417862_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10554960 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10565577 | - | - |
Mouse Genome 430 2.0 Array | 1442800_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1417862_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1417862_at | - | - |
Mouse Genome 430 2.0 Array | 1442800_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58238_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58238_PM_at | - | - |
Mouse Expression Set 430 | 1417862_PM_at | - | - |
Mouse Expression Set 430 | 1417862_at | - | - |
Mouse Expression Set 430 | 1442800_PM_x_at | - | - |
Mouse Expression Set 430 | 1442800_x_at | - | - |
Murine Genome U74v2 Set | 167568_i_at | - | - |
Murine Genome U74v2 Set | 167568_PM_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1417862_at | - | - |
Mouse Genome 430A 2.0 Array | 1417862_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417862_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417862_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442800_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442800_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3120072 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3120072 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003120072 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2756184 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2756184 | - | - |
mouse-6 v1.1 (Illumina) | scl32373.1.68_9-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3120072 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32373.1.68_9-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3120072 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1234519 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234519 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32373.1.68_9-S | - | - |
