Assay Details
Gene Symbol: 4930554H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019764.1 | - | 3 - 4 | 1282 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
cholinergic receptor, nicotinic, alpha polypeptide 7 |
Chrna7 |
Acra7, alpha7, nAchR |
-216.1kb | Sense | Chr. 7 - 63098692 - 63212526 on NCBI Build GRCm38 |
Downstream | OTU domain containing 7A | Otud7a |
AJ430384, Otud7 |
11.5kb | Anti-sense | Chr. 7 - 63444745 - 63759905 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
