Assay Details
| Gene Symbol: Cyr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010516.2 | NP_034646.1 | 3 - 4 | 813 | 73 | |
| GenBank mRNA | AK156723.1 | - | 3 - 4 | 808 | 73 | |
| AK159452.1 | - | 3 - 4 | 805 | 73 | ||
| BC003205.1 | - | 3 - 4 | 798 | 73 | ||
| BC066019.1 | - | 3 - 4 | 793 | 5716887 | 73 | |
| M32490.1 | - | 3 - 4 | 809 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyr61
Genomic Map