Assay Details
| Gene Symbol: Bcl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009740.2 | NP_033870.1 | 1 - 2 | 252 | 80 | |
| GenBank mRNA | AF057701.1 | - | 1 - 2 | 136 | 80 | |
| AF100339.1 | - | 1 - 2 | 144 | 80 | ||
| AF127387.1 | - | 1 - 2 | 55 | 80 | ||
| AF134396.1 | - | 1 - 2 | 136 | 80 | ||
| AJ006289.1 | - | 1 - 2 | 133 | 80 | ||
| AK076082.1 | - | 1 - 2 | 164 | 80 | ||
| AK140179.1 | - | 1 - 2 | 124 | 80 | ||
| AK146236.1 | - | 1 - 2 | 158 | 80 | ||
| AK150883.1 | - | 1 - 2 | 153 | 80 | ||
| AK152563.1 | - | 1 - 2 | 153 | 80 | ||
| AK152847.1 | - | 1 - 2 | 149 | 80 | ||
| AK156890.1 | - | 1 - 2 | 206 | 80 | ||
| AK168084.1 | - | 1 - 2 | 158 | 80 | ||
| AK169126.1 | - | 1 - 2 | 159 | 80 | ||
| AK172158.1 | - | 1 - 2 | 163 | 80 | ||
| BC003703.1 | - | 1 - 2 | 172 | 80 | ||
| BC024379.1 | - | 1 - 2 | 130 | 4976147 | 80 | |
| BC172007.1 | - | 1 - 2 | 136 | 9054293 | 80 | |
| BC172009.1 | - | 1 - 2 | 136 | 9054295 | 80 | |
| BU707261.1 | - | 1 - 2 | 252 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bcl10
Genomic Map