Assay Details
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009740.2 | NP_033870.1 | 2 - 3 | 549 | 86 | |
GenBank mRNA | AF057701.1 | - | 2 - 3 | 433 | 86 | |
AF100339.1 | - | 2 - 3 | 441 | 86 | ||
AF127387.1 | - | 2 - 3 | 352 | 86 | ||
AF134396.1 | - | 2 - 3 | 433 | 86 | ||
AJ006289.1 | - | 2 - 3 | 430 | 86 | ||
AK076082.1 | - | 2 - 3 | 461 | 86 | ||
AK140179.1 | - | 2 - 3 | 421 | 86 | ||
AK146236.1 | - | 2 - 3 | 455 | 86 | ||
AK150883.1 | - | 2 - 3 | 450 | 86 | ||
AK152563.1 | - | 2 - 3 | 450 | 86 | ||
AK152847.1 | - | 2 - 3 | 446 | 86 | ||
AK156890.1 | - | 2 - 3 | 503 | 86 | ||
AK168084.1 | - | 2 - 3 | 455 | 86 | ||
AK169126.1 | - | 2 - 3 | 456 | 86 | ||
AK172158.1 | - | 2 - 3 | 460 | 86 | ||
BC003703.1 | - | 2 - 3 | 468 | 86 | ||
BC024379.1 | - | 2 - 3 | 427 | 4976147 | 86 | |
BC172007.1 | - | 2 - 3 | 433 | 9054293 | 86 | |
BC172009.1 | - | 2 - 3 | 433 | 9054295 | 86 | |
BU707261.1 | - | 2 - 3 | 549 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
