Assay Details
| Gene Symbol: Odf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162539.1 | NP_001156011.1 | 11 - 12 | 1394 | 131 | |
| NM_025714.4 | NP_079990.2 | 12 - 13 | 1389 | 131 | ||
| GenBank mRNA | AK014630.1 | - | 12 - 13 | 1391 | 131 | |
| BC020075.1 | - | 11 - 12 | 1363 | 3979963 | 131 | |
More Information
Set Membership: |
|

Odf2l
Genomic Map