Assay Details
| Gene Symbol: Ddah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026993.3 | NP_081269.1 | 2 - 3 | 579 | 156 | |
| GenBank mRNA | AK010430.1 | - | 2 - 3 | 545 | 156 | |
| AK143507.1 | - | 2 - 3 | 596 | 156 | ||
| AK148907.1 | - | 2 - 3 | 580 | 156 | ||
| AK149185.1 | - | 2 - 3 | 231 | 156 | ||
| BC034505.1 | - | 2 - 3 | 530 | 5134831 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddah1
Genomic Map