Assay Details
| Gene Symbol: Abhd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205181.1 | NP_001192110.1 | 3 - 4 | 807 | 99 | |
| NM_134076.2 | NP_598837.2 | 3 - 4 | 552 | 99 | ||
| GenBank mRNA | AK030250.1 | - | 3 - 4 | 748 | 99 | |
| AK049366.1 | - | 3 - 4 | 533 | 99 | ||
| AK152595.1 | - | 3 - 4 | 553 | 99 | ||
| AK161853.1 | - | 3 - 4 | 502 | 99 | ||
| BC003982.1 | - | 1 - 2 | 192 | 99 | ||
| BC017532.1 | - | 3 - 4 | 487 | 4482138 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd4
Genomic Map