Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200000275.mm.1 |
NM_153082.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000275.mm.1 |
NM_153082.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000275.mm.1 |
TC1200000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000272.mm.1 |
NM_153082.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000272.mm.1 |
NM_153082.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000272.mm.1 |
TC1200000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000269.mm.1 |
NM_153082.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000269.mm.1 |
NM_153082.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000269.mm.1 |
TC1200000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000271.mm.1 |
NM_153082.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000271.mm.1 |
NM_153082.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000271.mm.1 |
TC1200000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000273.mm.1 |
NM_153082.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000273.mm.1 |
NM_153082.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000273.mm.1 |
TC1200000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000274.mm.1 |
NM_153082.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000274.mm.1 |
NM_153082.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000274.mm.1 |
TC1200000018.mm.1 |
MTA TC |