Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009653.mm.1 |
NM_001082483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009653.mm.1 |
NM_001082483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009653.mm.1 |
TC1200001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009656.mm.1 |
NM_001082483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009656.mm.1 |
NM_001082483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009656.mm.1 |
TC1200001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009655.mm.1 |
NM_001082483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009655.mm.1 |
NM_001082483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009655.mm.1 |
TC1200001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009654.mm.1 |
NM_001082483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009654.mm.1 |
NM_001082483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009654.mm.1 |
TC1200001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009658.mm.1 |
NM_001082483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009658.mm.1 |
NM_001082483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009658.mm.1 |
TC1200001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009657.mm.1 |
NM_001082483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009657.mm.1 |
NM_001082483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009657.mm.1 |
TC1200001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009651.mm.1 |
NM_001082483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009651.mm.1 |
NM_001082483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009651.mm.1 |
TC1200001348.mm.1 |
MTA TC |