Assay Details
| Gene Symbol: Efr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082483.1 | NP_001075952.1 | 6 - 7 | 837 | 84 | |
| GenBank mRNA | AK129247.1 | - | 2 - 3 | 284 | 84 | |
| BC075650.1 | - | 5 - 6 | 594 | 84 | ||
| BC147643.1 | - | 6 - 7 | 837 | 9056633 | 84 | |
| BC157968.1 | - | 6 - 7 | 837 | 9088136 | 84 | |
More Information
Set Membership: |
|

Efr3b
Genomic Map