Assay Details
| Gene Symbol: Spata17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028848.3 | NP_083124.1 | 1 - 2 | 279 | 82 | |
| NM_029067.1 | NP_083343.1 | 1 - 2 | 279 | 82 | ||
| GenBank mRNA | AK050925.1 | - | 1 - 2 | 280 | 82 | |
| AY747687.1 | - | 1 - 2 | 254 | 82 | ||
| BC089520.1 | - | 1 - 2 | 254 | 6774024 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spata17
Genomic Map