Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100030794.mm.1 |
NM_029067.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030794.mm.1 |
NM_029067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030794.mm.1 |
TC0100003761.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030795.mm.1 |
NM_028848.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030795.mm.1 |
NM_028848.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030795.mm.1 |
NM_029067.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030795.mm.1 |
NM_029067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030795.mm.1 |
TC0100003761.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030793.mm.1 |
NM_029067.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030793.mm.1 |
NM_029067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030793.mm.1 |
TC0100003761.mm.1 |
MTA TC |