Assay Details
| Gene Symbol: Kctd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172650.2 | NP_766238.1 | 1 - 2 | 309 | 68 | |
| GenBank mRNA | AK034380.1 | - | 1 - 2 | 309 | 68 | |
| AK049362.1 | - | 1 - 2 | 307 | 68 | ||
| AK087865.1 | - | 1 - 2 | 310 | 68 | ||
| AK139700.1 | - | 1 - 2 | 309 | 68 | ||
| BC061458.1 | - | 1 - 2 | 297 | 68 | ||
| BC079581.1 | - | 1 - 2 | 296 | 5702630 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kctd3
Genomic Map