Assay Details
| Gene Symbol: Kctd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172650.2 | NP_766238.1 | 9 - 10 | 1047 | 81 | |
| GenBank mRNA | AK033491.1 | - | 9 - 10 | 1048 | 81 | |
| AK034380.1 | - | 9 - 10 | 1048 | 81 | ||
| AK049362.1 | - | 9 - 10 | 1045 | 81 | ||
| AK087865.1 | - | 9 - 10 | 1048 | 81 | ||
| AK139700.1 | - | 9 - 10 | 1048 | 81 | ||
| BC061458.1 | - | 9 - 10 | 1035 | 81 | ||
| BC079581.1 | - | 9 - 10 | 1034 | 5702630 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kctd3
Genomic Map