Assay Details
| Gene Symbol: Prcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028243.3 | NP_082519.1 | 7 - 8 | 1163 | 99 | |
| GenBank mRNA | AK011112.1 | - | 7 - 8 | 1142 | 99 | |
| AK051677.1 | - | 7 - 8 | 1120 | 99 | ||
| AK054462.1 | - | 7 - 8 | 1101 | 99 | ||
| AK166282.1 | - | 7 - 8 | 1120 | 99 | ||
| BC026424.1 | - | 7 - 8 | 959 | 99 | ||
| BC055022.1 | - | 7 - 8 | 1096 | 4017462 | 99 | |
| BC089575.1 | - | 7 - 8 | 1013 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prcp
Genomic Map