Assay Details
| Gene Symbol: Igsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177591.4 | NP_808259.2 | 5 - 6 | 813 | 61 | |
| NM_183335.2 | NP_899178.2 | 4 - 5 | 677 | 61 | ||
| GenBank mRNA | AK129126.1 | - | 5 - 6 | 795 | 61 | |
| AY227771.1 | - | 5 - 6 | 807 | 61 | ||
| AY227773.1 | - | 4 - 5 | 677 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igsf1
Genomic Map