Assay Details
| Gene Symbol: Dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190717.1 | NP_001177646.1 | 2 - 3 | 424 | 138 | |
| NM_013726.3 | NP_038754.1 | 2 - 3 | 424 | 138 | ||
| GenBank mRNA | AJ003132.1 | - | 2 - 3 | 314 | 138 | |
| AK014480.1 | - | 2 - 3 | 390 | 138 | ||
| AK132403.1 | - | 2 - 3 | 389 | 138 | ||
| BB611145.1 | - | 2 - 3 | 425 | 138 | ||
| BC069983.1 | - | 2 - 3 | 372 | 138 | ||
| BC138709.1 | - | 2 - 3 | 242 | 8861731 | 138 | |
| BC138710.1 | - | 2 - 3 | 242 | 8861732 | 138 | |
| BC145392.1 | - | 2 - 3 | 242 | 9053950 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dbf4
Genomic Map