Assay Details
| Gene Symbol: Dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013726.3 | NP_038754.1 | 7 - 8 | 837 | 134 | |
| GenBank mRNA | AJ003132.1 | - | 7 - 8 | 727 | 134 | |
| AK014480.1 | - | 7 - 8 | 803 | 134 | ||
| AK083650.1 | - | 4 - 5 | 212 | 134 | ||
| AK163408.1 | - | 5 - 6 | 413 | 134 | ||
| BC069983.1 | - | 7 - 8 | 785 | 134 | ||
| BC138709.1 | - | 7 - 8 | 655 | 8861731 | 134 | |
| BC138710.1 | - | 7 - 8 | 655 | 8861732 | 134 | |
More Information
Set Membership: |
|

Dbf4
Genomic Map