Assay Details
| Gene Symbol: Igfl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003393.1 | NP_001003393.1 | 1 - 2 | 52 | 92 | |
| GenBank mRNA | AY672115.1 | - | 1 - 2 | 52 | 92 | |
| BC147097.1 | - | 1 - 2 | 52 | 9056138 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY672115.1, BC147097.1, NM_001003393.1 transcripts.
Set Membership: |
|

Igfl3
Genomic Map