Assay Details
| Gene Symbol: Arhgap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172525.2 | NP_766113.1 | 4 - 5 | 554 | 86 | |
| GenBank mRNA | AK043637.1 | - | 4 - 5 | 601 | 86 | |
| AK044979.1 | - | 4 - 5 | 604 | 86 | ||
| AK139908.1 | - | 2 - 3 | 511 | 86 | ||
| BC040387.1 | - | 4 - 5 | 578 | 3599962 | 86 | |
| BC052858.1 | - | 4 - 5 | 578 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap29
Genomic Map