Assay Details
| Gene Symbol: Pomt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145145.1 | NP_660127.1 | 13 - 14 | 1361 | 57 | |
| GenBank mRNA | AK053889.1 | - | 12 - 13 | 1881 | 57 | |
| AK134770.1 | - | 13 - 14 | 1405 | 57 | ||
| AK153984.1 | - | 13 - 14 | 1460 | 57 | ||
| AY007238.1 | - | 13 - 14 | 1401 | 57 | ||
| AY494857.1 | - | 13 - 14 | 1421 | 57 | ||
| BC027325.1 | - | 13 - 14 | 1361 | 3981366 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pomt1
Genomic Map