Assay Details
| Gene Symbol: Lcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008491.1 | NP_032517.1 | 3 - 4 | 380 | 65 | |
| GenBank mRNA | AK002932.1 | - | 3 - 4 | 413 | 65 | |
| AK149774.1 | - | 3 - 4 | 412 | 65 | ||
| BC020275.1 | - | 3 - 4 | 380 | 65 | ||
| BC132069.1 | - | 3 - 4 | 380 | 40130346 | 65 | |
| BC132071.1 | - | 3 - 4 | 380 | 40130348 | 65 | |
| S82469.1 | - | 3 - 4 | 344 | 72 | ||
| X14607.1 | - | 3 - 4 | 380 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lcn2
Genomic Map