Assay Details
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301737.1 | NP_001288666.1 | 15 - 16 | 1784 | 67 | |
| NM_010065.3 | NP_034195.2 | 14 - 15 | 1772 | 67 | ||
| NR_125959.1 | 15 - 16 | 1784 | 67 | |||
| GenBank mRNA | AK144142.1 | - | 14 - 15 | 1723 | 67 | |
| AK220483.1 | - | 15 - 16 | 1726 | 67 | ||
| BC034679.1 | - | 15 - 16 | 1738 | 4506865 | 67 | |
| BC058623.1 | - | 15 - 16 | 1718 | 5706803 | 67 | |
| BM950315.1 | - | 1 - 2 | 177 | 67 | ||
| BQ179370.1 | - | 9 - 10 | 709 | 67 | ||
| CO425806.1 | - | 1 - 2 | 103 | 67 | ||
| L31395.1 | - | 15 - 16 | 1681 | 67 | ||
| L31396.1 | - | 10 - 11 | 953 | 67 | ||
| L31397.1 | - | 14 - 15 | 1725 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnm1
Genomic Map