Assay Details
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301737.1 | NP_001288666.1 | 3 - 4 | 506 | 59 | |
| NM_010065.3 | NP_034195.2 | 3 - 4 | 506 | 59 | ||
| NR_125959.1 | 3 - 4 | 506 | 59 | |||
| GenBank mRNA | AK011651.1 | - | 3 - 4 | 473 | 59 | |
| AK144142.1 | - | 3 - 4 | 457 | 59 | ||
| AK220483.1 | - | 3 - 4 | 448 | 59 | ||
| BC034679.1 | - | 3 - 4 | 460 | 4506865 | 59 | |
| BC058623.1 | - | 3 - 4 | 440 | 5706803 | 59 | |
| CA328097.1 | - | 1 - 2 | 132 | 59 | ||
| L31397.1 | - | 3 - 4 | 459 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnm1
Genomic Map