Assay Details
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011836.3 | NP_035966.2 | 19 - 20 | 3513 | 56 | |
| GenBank mRNA | AF083372.1 | - | 19 - 20 | 3450 | 56 | |
| AK132948.1 | - | 16 - 17 | 2500 | 56 | ||
| BC005535.1 | - | 2 - 3 | 258 | 56 | ||
| BC096366.1 | - | 19 - 20 | 3513 | 2865309 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lamc3
Genomic Map