Assay Details
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011836.3 | NP_035966.2 | 1 - 2 | 466 | 69 | |
| GenBank mRNA | AF079520.1 | - | 1 - 2 | 456 | 69 | |
| BC096366.1 | - | 1 - 2 | 466 | 2865309 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lamc3
Genomic Map