Assay Details
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011836.3 | NP_035966.2 | 20 - 21 | 3586 | 67 | |
| GenBank mRNA | AF079520.1 | - | 20 - 21 | 3609 | 67 | |
| AF083372.1 | - | 20 - 21 | 3523 | 67 | ||
| AK132948.1 | - | 17 - 18 | 2573 | 67 | ||
| BC005535.1 | - | 3 - 4 | 331 | 67 | ||
| BC096366.1 | - | 20 - 21 | 3586 | 2865309 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lamc3
Genomic Map