Assay Details
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011836.3 | NP_035966.2 | 7 - 8 | 1473 | 90 | |
| GenBank mRNA | AF083372.1 | - | 7 - 8 | 1410 | 90 | |
| AK132948.1 | - | 4 - 5 | 458 | 90 | ||
| BC096366.1 | - | 7 - 8 | 1473 | 2865309 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lamc3
Genomic Map