Assay Details
Gene Symbol: Armc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081083.2 | NP_001074552.1 | 4 - 5 | 530 | 98 | |
NM_001271563.1 | NP_001258492.1 | 4 - 5 | 530 | 98 | ||
NM_001271564.1 | NP_001258493.1 | 4 - 5 | 530 | 98 | ||
NM_001271565.1 | NP_001258494.1 | 4 - 5 | 530 | 98 | ||
GenBank mRNA | AK014880.1 | - | 4 - 5 | 346 | 98 | |
AK041744.1 | - | 4 - 5 | 354 | 98 | ||
BC145004.1 | - | 4 - 5 | 338 | 98 | ||
BC145005.1 | - | 4 - 5 | 338 | 9053553 | 98 | |
BC145013.1 | - | 4 - 5 | 338 | 9053561 | 98 | |
BC150688.1 | - | 4 - 5 | 338 | 9087599 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
