Assay Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290800.1 | NP_001277729.1 | 15 - 16 | 2582 | 79 | |
| NM_019392.2 | NP_062265.2 | 15 - 16 | 2118 | 79 | ||
| GenBank mRNA | AB000828.1 | - | 15 - 16 | 1855 | 79 | |
| AK141198.1 | - | 15 - 16 | 1889 | 79 | ||
| BC066058.1 | - | 15 - 16 | 2118 | 6834015 | 79 | |
| BC082325.1 | - | 15 - 16 | 2066 | 30356080 | 79 | |
| U05683.1 | - | 15 - 16 | 1903 | 79 | ||
| U18342.1 | - | 15 - 16 | 2035 | 79 | ||
| U18343.1 | - | 15 - 16 | 2578 | 79 | ||
| U18933.1 | - | 15 - 16 | 2054 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyro3
Genomic Map