Assay Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290800.1 | NP_001277729.1 | 3 - 4 | 1121 | 101 | |
| NM_019392.2 | NP_062265.2 | 3 - 4 | 657 | 101 | ||
| GenBank mRNA | AB000828.1 | - | 3 - 4 | 394 | 101 | |
| AK141198.1 | - | 3 - 4 | 428 | 101 | ||
| BC066058.1 | - | 3 - 4 | 657 | 6834015 | 101 | |
| BC082325.1 | - | 3 - 4 | 605 | 30356080 | 101 | |
| D17393.1 | - | 3 - 4 | 1102 | 101 | ||
| U05683.1 | - | 3 - 4 | 442 | 101 | ||
| U18342.1 | - | 3 - 4 | 574 | 101 | ||
| U18343.1 | - | 3 - 4 | 1117 | 101 | ||
| U18933.1 | - | 3 - 4 | 593 | 101 | ||
| X78103.1 | - | 3 - 4 | 601 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyro3
Genomic Map