Assay Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290800.1 | NP_001277729.1 | 8 - 9 | 1817 | 80 | |
| NM_019392.2 | NP_062265.2 | 8 - 9 | 1353 | 80 | ||
| GenBank mRNA | AB000828.1 | - | 8 - 9 | 1090 | 80 | |
| AK141198.1 | - | 8 - 9 | 1124 | 80 | ||
| BC066058.1 | - | 8 - 9 | 1353 | 6834015 | 80 | |
| BC082325.1 | - | 8 - 9 | 1301 | 30356080 | 80 | |
| D17393.1 | - | 8 - 9 | 1795 | 80 | ||
| U05683.1 | - | 8 - 9 | 1138 | 80 | ||
| U18342.1 | - | 8 - 9 | 1270 | 80 | ||
| U18343.1 | - | 8 - 9 | 1813 | 80 | ||
| U18933.1 | - | 8 - 9 | 1289 | 80 | ||
| X78103.1 | - | 8 - 9 | 1297 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyro3
Genomic Map