Assay Details
| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027175.3 | NP_081451.3 | 2 - 3 | 847 | 71 | |
| GenBank mRNA | AK010328.1 | - | 1 - 2 | 677 | 71 | |
| AK050343.1 | - | 2 - 3 | 848 | 71 | ||
| BC018422.1 | - | 2 - 3 | 677 | 3593650 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufaf1
Genomic Map