Assay Details
| Gene Symbol: Ltk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008523.2 | NP_032549.2 | 10 - 11 | 1302 | 85 | |
| NM_203345.2 | NP_976220.2 | 16 - 17 | 2148 | 85 | ||
| NM_206941.1 | NP_996824.1 | 11 - 12 | 1485 | 85 | ||
| NM_206942.2 | NP_996825.2 | 15 - 16 | 1965 | 85 | ||
| GenBank mRNA | AK157478.1 | - | 8 - 9 | 1299 | 85 | |
| BC047138.1 | - | 10 - 11 | 1302 | 6389834 | 85 | |
| BC055898.1 | - | 9 - 10 | 1166 | 85 | ||
| BC141299.1 | - | 15 - 16 | 1941 | 9055911 | 85 | |
| X07984.1 | - | 10 - 11 | 1276 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ltk
Genomic Map