Assay Details
| Gene Symbol: Ltk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008523.2 | NP_032549.2 | 11 - 12 | 1400 | 78 | |
| NM_203345.2 | NP_976220.2 | 17 - 18 | 2246 | 78 | ||
| NM_206941.1 | NP_996824.1 | 12 - 13 | 1583 | 78 | ||
| NM_206942.2 | NP_996825.2 | 16 - 17 | 2063 | 78 | ||
| GenBank mRNA | AK157478.1 | - | 9 - 10 | 1397 | 78 | |
| BC047138.1 | - | 11 - 12 | 1400 | 6389834 | 78 | |
| BC141299.1 | - | 16 - 17 | 2039 | 9055911 | 78 | |
| M90470.1 | - | 17 - 18 | 2246 | 78 | ||
| X07984.1 | - | 11 - 12 | 1374 | 78 | ||
| X52621.1 | - | 11 - 12 | 1389 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ltk
Genomic Map