Assay Details
| Gene Symbol: Ltk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008523.2 | NP_032549.2 | 5 - 6 | 825 | 67 | |
| NM_203345.2 | NP_976220.2 | 11 - 12 | 1671 | 67 | ||
| NM_206941.1 | NP_996824.1 | 6 - 7 | 1008 | 67 | ||
| NM_206942.2 | NP_996825.2 | 10 - 11 | 1488 | 67 | ||
| GenBank mRNA | AK157478.1 | - | 4 - 5 | 639 | 67 | |
| BC047138.1 | - | 5 - 6 | 825 | 6389834 | 67 | |
| BC055898.1 | - | 5 - 6 | 819 | 67 | ||
| BC141299.1 | - | 10 - 11 | 1464 | 9055911 | 67 | |
| M90470.1 | - | 11 - 12 | 1671 | 67 | ||
| X07984.1 | - | 5 - 6 | 799 | 67 | ||
| X52621.1 | - | 5 - 6 | 814 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ltk
Genomic Map