Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200037315.mm.1 |
NM_133838.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037315.mm.1 |
NM_133838.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037315.mm.1 |
TC0200004454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037316.mm.1 |
NM_133838.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037316.mm.1 |
NM_133838.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037316.mm.1 |
TC0200004454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037313.mm.1 |
NM_133838.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037313.mm.1 |
NM_133838.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037313.mm.1 |
TC0200004454.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037317.mm.1 |
NM_133838.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037317.mm.1 |
NM_133838.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037317.mm.1 |
TC0200004454.mm.1 |
MTA TC |