Assay Details
| Gene Symbol: Cfap61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175280.3 | NP_780489.2 | 4 - 5 | 432 | 51 | |
| GenBank mRNA | AK019712.1 | - | 433 | 51 | ||
| AK038772.1 | - | 4 - 5 | 433 | 51 | ||
| BC057361.1 | - | 4 - 5 | 400 | 51 | ||
| BC058733.1 | - | 4 - 5 | 400 | 51 | ||
| BC107030.1 | - | 4 - 5 | 418 | 40050496 | 51 | |
| BC107031.1 | - | 4 - 5 | 413 | 40050497 | 51 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cfap61
Genomic Map