Assay Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021526.2 | NP_067501.2 | 1 - 2 | 180 | 134 | |
| GenBank mRNA | AK011974.1 | - | 1 - 2 | 129 | 134 | |
| AK012013.1 | - | 1 - 2 | 111 | 134 | ||
| AK031863.1 | - | 1 - 2 | 118 | 134 | ||
| AK078134.1 | - | 1 - 2 | 131 | 134 | ||
| AK136306.1 | - | 1 - 2 | 116 | 134 | ||
| AK150280.1 | - | 1 - 2 | 116 | 134 | ||
| AK172694.1 | - | 1 - 2 | 119 | 134 | ||
| BC003742.1 | - | 1 - 2 | 85 | 3599671 | 134 | |
| BY324928.1 | - | 1 - 2 | 182 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011974.1, AK012013.1, AK150280.1, BC003742.1, NM_021526.2 transcripts.
Set Membership: |
|

Psmd14
Genomic Map