Assay Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021526.2 | NP_067501.2 | 2 - 3 | 301 | 168 | |
| GenBank mRNA | AK011974.1 | - | 2 - 3 | 250 | 168 | |
| AK012013.1 | - | 2 - 3 | 232 | 168 | ||
| AK031863.1 | - | 2 - 3 | 239 | 168 | ||
| AK078134.1 | - | 2 - 3 | 252 | 168 | ||
| AK150280.1 | - | 2 - 3 | 237 | 168 | ||
| AK151104.1 | - | 2 - 3 | 284 | 168 | ||
| AK151733.1 | - | 2 - 3 | 284 | 168 | ||
| BC003742.1 | - | 2 - 3 | 206 | 3599671 | 168 | |
| BY324928.1 | - | 2 - 3 | 303 | 168 | ||
| Y13071.1 | - | 2 - 3 | 225 | 168 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011974.1, AK012013.1, AK150280.1, AK151104.1, AK151733.1, BC003742.1, NM_021526.2, Y13071.1 transcripts.
Set Membership: |
|

Psmd14
Genomic Map