Assay Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021526.2 | NP_067501.2 | 3 - 4 | 362 | 67 | |
| GenBank mRNA | AK011974.1 | - | 3 - 4 | 311 | 67 | |
| AK012013.1 | - | 3 - 4 | 293 | 67 | ||
| AK031863.1 | - | 3 - 4 | 300 | 67 | ||
| AK078134.1 | - | 3 - 4 | 313 | 67 | ||
| AK150280.1 | - | 3 - 4 | 298 | 67 | ||
| AK151104.1 | - | 3 - 4 | 345 | 67 | ||
| AK151733.1 | - | 3 - 4 | 345 | 67 | ||
| BC003742.1 | - | 3 - 4 | 267 | 3599671 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmd14
Genomic Map