Assay Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162503.1 | NP_001155975.1 | 3 - 4 | 499 | 152 | |
| NM_021507.5 | NP_067482.4 | 3 - 4 | 818 | 152 | ||
| NR_027888.1 | 3 - 4 | 454 | 152 | |||
| GenBank mRNA | AF174535.1 | - | 3 - 4 | 818 | 152 | |
| AK016166.1 | - | 3 - 4 | 451 | 152 | ||
| AK075598.1 | - | 3 - 4 | 500 | 152 | ||
| AK149948.1 | - | 3 - 4 | 470 | 152 | ||
| BC011153.1 | - | 2 - 3 | 437 | 4196280 | 152 | |
| BC028247.1 | - | 3 - 4 | 487 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sqrdl
Genomic Map