Assay Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162503.1 | NP_001155975.1 | 5 - 6 | 759 | 85 | |
| NM_021507.5 | NP_067482.4 | 5 - 6 | 1078 | 85 | ||
| NR_027888.1 | 5 - 6 | 714 | 85 | |||
| GenBank mRNA | AF174535.1 | - | 5 - 6 | 1078 | 85 | |
| AK008290.1 | - | 5 - 6 | 725 | 85 | ||
| AK016166.1 | - | 5 - 6 | 711 | 86 | ||
| AK075598.1 | - | 5 - 6 | 760 | 85 | ||
| AK149948.1 | - | 5 - 6 | 730 | 85 | ||
| BC011153.1 | - | 4 - 5 | 697 | 4196280 | 85 | |
| BC028247.1 | - | 5 - 6 | 747 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sqrdl
Genomic Map