Assay Details
| Gene Symbol: Nedd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111324.2 | NP_001104794.1 | 3 - 4 | 811 | 58 | |
| NM_017464.5 | NP_059492.3 | 2 - 3 | 592 | 58 | ||
| GenBank mRNA | AK030985.1 | - | 3 - 4 | 745 | 58 | |
| AK033729.1 | - | 2 - 3 | 593 | 58 | ||
| AK054179.1 | - | 2 - 3 | 588 | 58 | ||
| AK083374.1 | - | 3 - 4 | 739 | 58 | ||
| BC053713.1 | - | 3 - 4 | 744 | 30077589 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nedd9
Genomic Map